siRNA Details
| Gene Symbol: Epn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033914.1 | NP_001029086.1 | 8 | 1626 | ||
| NM_021852.2 | NP_068624.2 | 7 | 1455 | |||
| XM_006246521.2 | XP_006246583.1 | 8 | 1410 | |||
| XM_006246522.2 | XP_006246584.1 | 9 | 1708 | |||
| XM_006246523.2 | XP_006246585.1 | 7 | 1239 | |||
| GenBank mRNA | AF096269.1 | - | 6 | 1196 | ||
| BC094524.1 | - | 8 | 1605 | 7318094 | ||
| FQ211574.1 | - | 9 | 1708 | |||
| FQ211939.1 | - | 8 | 1650 | |||

Epn2
Genomic Map