siRNA Details
| Gene Symbol: C14orf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007176.3 | NP_009107.1 | 5 | 1375 | ||
| GenBank mRNA | AF134159.2 | - | 5 | 995 | ||
| AF136971.1 | - | 5 | 1035 | |||
| BC002444.2 | - | 5 | 1040 | 3347242 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C14orf1| Gene Symbol: C14orf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007176.3 | NP_009107.1 | 5 | 1375 | ||
| GenBank mRNA | AF134159.2 | - | 5 | 995 | ||
| AF136971.1 | - | 5 | 1035 | |||
| BC002444.2 | - | 5 | 1040 | 3347242 | ||