siRNA Details
| Gene Symbol: Crx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021855.1 | NP_068627.1 | 3 | 387 | ||
| XM_006228354.1 | XP_006228416.1 | 4 | 571 | |||
| GenBank mRNA | AB021129.1 | - | 3 | 387 | ||
| AF135838.1 | - | 3 | 387 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crx| Gene Symbol: Crx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021855.1 | NP_068627.1 | 3 | 387 | ||
| XM_006228354.1 | XP_006228416.1 | 4 | 571 | |||
| GenBank mRNA | AB021129.1 | - | 3 | 387 | ||
| AF135838.1 | - | 3 | 387 | |||