siRNA Details
| Gene Symbol: Hspa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021863.3 | NP_068635.1 | 1 | 886 | ||
| XM_006240247.2 | XP_006240309.1 | 2 | 863 | |||
| GenBank mRNA | BC081803.1 | - | 2 | 857 | 7112083 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008301, NM_001002012, AK132261, BC052350, BC004714, and others.

Hspa2
Genomic Map