siRNA Details
| Gene Symbol: Fxyd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270688.1 | NP_001257617.1 | 5 | 255 | ||
| NM_001270689.1 | NP_001257618.1 | 5 | 283 | |||
| NM_021909.2 | NP_068709.2 | 5 | 359 | |||
| GenBank mRNA | BC060571.1 | - | 5 | 247 | 6891214 | |
| DV726876.1 | - | 1 | 25 | |||
| FM122563.1 | - | 5 | 233 | |||
| FQ220058.1 | - | 5 | 283 | |||

Fxyd5
Genomic Map