siRNA Details
| Gene Symbol: LOC60591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022203.1 | NP_071539.1 | 2 | 170 | ||
| GenBank mRNA | U79661.1 | - | 2 | 170 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC60591| Gene Symbol: LOC60591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022203.1 | NP_071539.1 | 2 | 170 | ||
| GenBank mRNA | U79661.1 | - | 2 | 170 | ||