siRNA Details
| Gene Symbol: Ppp2r2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022209.3 | NP_071545.2 | 2, 3 | 369 | ||
| GenBank mRNA | AY251277.1 | - | 2, 3 | 321 | ||
| BC078834.1 | - | 5, 6 | 578 | 7114294 | ||
| D14421.1 | - | 2, 3 | 144 | |||
| D38260.1 | - | 2, 3 | 164 | |||
| FM080850.1 | - | 2, 3 | 369 | |||

Ppp2r2b
Genomic Map