siRNA Details
| Gene Symbol: Pik3r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022213.1 | NP_071549.1 | 3 | 299 | ||
| XM_006238714.2 | XP_006238776.1 | 4 | 320 | |||
| XM_008763979.1 | XP_008762201.1 | 4 | 522 | |||
| GenBank mRNA | D64047.1 | - | 3 | 299 | ||

Pik3r3
Genomic Map