siRNA Details
| Gene Symbol: Dnajc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022232.1 | NP_071568.1 | 2 | 125 | ||
| GenBank mRNA | AB017702.3 | - | 2 | 125 | ||
| BC061764.1 | - | 2 | 220 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajc3| Gene Symbol: Dnajc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022232.1 | NP_071568.1 | 2 | 125 | ||
| GenBank mRNA | AB017702.3 | - | 2 | 125 | ||
| BC061764.1 | - | 2 | 220 | |||