siRNA Details
| Gene Symbol: Slc33a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022252.2 | NP_071588.2 | 1 | 372 | ||
| XM_008761058.1 | XP_008759280.1 | 1 | 796 | |||
| GenBank mRNA | BC078832.1 | - | 1 | 372 | 7132173 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc33a1| Gene Symbol: Slc33a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022252.2 | NP_071588.2 | 1 | 372 | ||
| XM_008761058.1 | XP_008759280.1 | 1 | 796 | |||
| GenBank mRNA | BC078832.1 | - | 1 | 372 | 7132173 | |