siRNA Details
| Gene Symbol: Bspry | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022261.2 | NP_071597.2 | 3 | 690 | ||
| GenBank mRNA | AF245225.1 | - | 1 | 217 | ||
| BC062051.1 | - | 3 | 638 | |||
| BC095901.1 | - | 3 | 341 | |||
| BC111380.1 | - | 2 | 244 | |||
| BC166587.1 | - | 3 | 690 | 7937509 | ||

Bspry
Genomic Map