siRNA Details
| Gene Symbol: Bspry | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022261.2 | NP_071597.2 | 5 | 792 | ||
| GenBank mRNA | AF245225.1 | - | 3 | 319 | ||
| BC062051.1 | - | 5 | 740 | |||
| BC095901.1 | - | 5 | 443 | |||
| BC111380.1 | - | 4 | 346 | |||
| BC166587.1 | - | 5 | 792 | 7937509 | ||

Bspry
Genomic Map