siRNA Details
Gene Symbol: Slc22a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022270.1 | NP_071606.1 | 651 | |||
GenBank mRNA | AF169831.1 | - | 651 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc22a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022270.1 | NP_071606.1 | 651 | |||
GenBank mRNA | AF169831.1 | - | 651 |