siRNA Details
| Gene Symbol: Slc22a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022270.1 | NP_071606.1 | 2213 | |||
| GenBank mRNA | AF169831.1 | - | 2213 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc22a4| Gene Symbol: Slc22a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022270.1 | NP_071606.1 | 2213 | |||
| GenBank mRNA | AF169831.1 | - | 2213 | |||