siRNA Details
| Gene Symbol: Casp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022277.1 | NP_071613.1 | 2 | 552 | ||
| XM_006245014.2 | XP_006245076.1 | 2 | 534 | |||
| XM_006245015.2 | XP_006245077.1 | 2 | 675 | |||
| XM_008767135.1 | XP_008765357.1 | 2 | 534 | |||
| GenBank mRNA | AF279308.1 | - | 2 | 552 | ||
| AF288372.1 | - | 2 | 287 | |||
| FQ226407.1 | - | 2 | 566 | |||

Casp8
Genomic Map