siRNA Details
| Gene Symbol: Eltd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022294.1 | NP_071630.1 | 2, 3 | 401 | ||
| GenBank mRNA | AF192401.1 | - | 2, 3 | 401 | ||
| AF192402.1 | - | 2, 3 | 401 | |||
More Information
Set Membership: |
|

Eltd1
Genomic Map