siRNA Details
Gene Symbol: Pi4ka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022301.1 | NP_071637.1 | 8 | 835 | ||
XM_008768902.1 | XP_008767124.1 | 8 | 979 | |||
GenBank mRNA | D83538.1 | - | 8 | 1113 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pi4ka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022301.1 | NP_071637.1 | 8 | 835 | ||
XM_008768902.1 | XP_008767124.1 | 8 | 979 | |||
GenBank mRNA | D83538.1 | - | 8 | 1113 |