siRNA Details
| Gene Symbol: Card9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022303.1 | NP_071639.1 | 4 | 657 | ||
| XM_008761625.1 | XP_008759847.1 | 4 | 1845 | |||
| XM_008761626.1 | XP_008759848.1 | 4 | 1845 | |||
| GenBank mRNA | AF311288.1 | - | 4 | 657 | ||

Card9
Genomic Map