siRNA Details
| Gene Symbol: Clec10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022393.2 | NP_071788.2 | 7 | 753 | ||
| XM_006246803.2 | XP_006246865.1 | 7 | 791 | |||
| XM_008767867.1 | XP_008766089.1 | 7 | 791 | |||
| XM_008767868.1 | XP_008766090.1 | 7 | 635 | |||
| XM_008767869.1 | XP_008766091.1 | 7 | 632 | |||
| GenBank mRNA | BC091151.1 | - | 7 | 753 | 7375234 | |
| FQ213042.1 | - | 7 | 625 | |||
| FQ221204.1 | - | 7 | 642 | |||
| FQ228437.1 | - | 7 | 611 | |||
| J05495.1 | - | 7 | 613 | |||

Clec10a
Genomic Map