siRNA Details
| Gene Symbol: Hnrnpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037285.1 | NP_001032362.1 | 4 | 832 | ||
| NM_001037286.1 | NP_001032363.1 | 3 | 874 | |||
| NM_001037287.1 | NP_001032364.1 | 4 | 789 | |||
| NM_022397.3 | NP_071792.1 | 4 | 803 | |||
| XM_008763262.1 | XP_008761484.1 | 1 | 561 | |||
| GenBank mRNA | AB022209.1 | - | 1 | 526 | ||
| BC097275.1 | - | 4 | 772 | 7444983 | ||
| BC103634.1 | - | 4 | 791 | 7097633 | ||

Hnrnpf
Genomic Map