siRNA Details
| Gene Symbol: Rpl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022510.1 | NP_071955.1 | 4, 5 | 480 | ||
| GenBank mRNA | X82180.1 | - | 4, 5 | 480 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpl4| Gene Symbol: Rpl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022510.1 | NP_071955.1 | 4, 5 | 480 | ||
| GenBank mRNA | X82180.1 | - | 4, 5 | 480 | ||