siRNA Details
| Gene Symbol: Sult1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022513.2 | NP_071958.1 | 1 | 202 | ||
| GenBank mRNA | D89375.1 | - | 1 | 153 | ||
| FQ209476.1 | - | 1 | 171 | |||
| FQ209486.1 | - | 1 | 236 | |||
| FQ209579.1 | - | 1 | 257 | |||
| FQ212608.1 | - | 1 | 213 | |||
| FQ218295.1 | - | 1 | 215 | |||
| U38419.1 | - | 1 | 205 | |||

Sult1b1
Genomic Map