siRNA Details
| Gene Symbol: Sult1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022513.2 | NP_071958.1 | 2 | 308 | ||
| XM_008770056.1 | XP_008768278.1 | 2 | 182 | |||
| GenBank mRNA | D89375.1 | - | 2 | 259 | ||
| FQ209476.1 | - | 2 | 277 | |||
| FQ209486.1 | - | 2 | 342 | |||
| FQ209579.1 | - | 2 | 363 | |||
| FQ210801.1 | - | 2 | 350 | |||
| FQ212608.1 | - | 2 | 319 | |||
| FQ218295.1 | - | 2 | 321 | |||
| U38419.1 | - | 2 | 311 | |||

Sult1b1
Genomic Map