siRNA Details
| Gene Symbol: Dap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022526.2 | NP_071971.1 | 4 | 883 | ||
| GenBank mRNA | FM053394.1 | - | 1 | 215 | ||
| FQ218203.1 | - | 4 | 768 | |||
| FQ228698.1 | - | 4 | 721 | |||
| FQ228865.1 | - | 4 | 727 | |||
| U05334.1 | - | 4 | 887 | |||

Dap
Genomic Map