siRNA Details
| Gene Symbol: Pllp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022533.1 | NP_071978.1 | 4 | 1076 | ||
| GenBank mRNA | BC078823.1 | - | 4 | 956 | 7108239 | |
| U13617.1 | - | 4 | 849 | |||
| Z49858.1 | - | 4 | 1076 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pllp| Gene Symbol: Pllp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022533.1 | NP_071978.1 | 4 | 1076 | ||
| GenBank mRNA | BC078823.1 | - | 4 | 956 | 7108239 | |
| U13617.1 | - | 4 | 849 | |||
| Z49858.1 | - | 4 | 1076 | |||