siRNA Details
Gene Symbol: Timm8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022541.2 | NP_071986.1 | 2 | 298 | ||
GenBank mRNA | AF196315.1 | - | 2 | 299 | ||
CF977529.1 | - | 2 | 308 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Timm8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022541.2 | NP_071986.1 | 2 | 298 | ||
GenBank mRNA | AF196315.1 | - | 2 | 299 | ||
CF977529.1 | - | 2 | 308 |