siRNA Details
| Gene Symbol: Timm8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022541.2 | NP_071986.1 | 2 | 466 | ||
| GenBank mRNA | AF196315.1 | - | 2 | 467 | ||
| CF977529.1 | - | 2 | 476 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Timm8b| Gene Symbol: Timm8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022541.2 | NP_071986.1 | 2 | 466 | ||
| GenBank mRNA | AF196315.1 | - | 2 | 467 | ||
| CF977529.1 | - | 2 | 476 | |||