siRNA Details
| Gene Symbol: Dapk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022546.1 | NP_071991.1 | 4 | 582 | ||
| XM_006240991.1 | XP_006241053.1 | 4 | 713 | |||
| XM_006240993.2 | XP_006241055.1 | 4 | 820 | |||
| XM_008765129.1 | XP_008763351.1 | 4 | 1046 | |||
| GenBank mRNA | AB010083.1 | - | 1 | 61 | ||
| AJ006971.1 | - | 4 | 582 | |||
| BC062076.1 | - | 4 | 583 | 5599435 | ||

Dapk3
Genomic Map