siRNA Details
Gene Symbol: Golga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022596.2 | NP_072118.2 | 26 | 4217 | ||
GenBank mRNA | U35022.2 | - | 26 | 4220 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Golga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022596.2 | NP_072118.2 | 26 | 4217 | ||
GenBank mRNA | U35022.2 | - | 26 | 4220 |