siRNA Details
| Gene Symbol: Ndufv3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101011.2 | NP_001094481.1 | 2 | 237 | ||
| NM_022607.2 | NP_072129.2 | 2 | 237 | |||
| GenBank mRNA | AB000098.1 | - | 2 | 168 | ||
| BC059134.1 | - | 193 | ||||
| CK365764.1 | - | 2 | 248 | |||
| DV725422.1 | - | 2 | 235 | |||
| FQ216920.1 | - | 2 | 185 | |||
| FQ217920.1 | - | 2 | 185 | |||
| FQ218041.1 | - | 2 | 185 | |||
| FQ223657.1 | - | 2 | 185 | |||

Ndufv3
Genomic Map