siRNA Details
| Gene Symbol: Sept7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113740.1 | NP_001107212.1 | 3 | 203 | ||
| NM_022616.2 | NP_072138.2 | 2 | 200 | |||
| GenBank mRNA | AF142759.1 | - | 2 | 167 | ||
| BC128738.1 | - | 3 | 101 | 7381331 | ||
| CA512303.1 | - | 3 | 204 | |||
| EV764168.1 | - | 3 | 175 | |||
| Gene Symbol: LOC100910754 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772553.1 | XP_008770775.1 | 1 | 192 | ||

Sept7
Genomic Map