siRNA Details
Gene Symbol: Tpc1808 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022625.2 | NP_072147.1 | 1408 | |||
GenBank mRNA | AF078811.2 | - | 1408 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tpc1808 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022625.2 | NP_072147.1 | 1408 | |||
GenBank mRNA | AF078811.2 | - | 1408 |