siRNA Details
| Gene Symbol: Tmprss11d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033652.1 | NP_001028824.1 | 7 | 785 | ||
| NM_022630.1 | NP_072152.1 | 2 | 156 | |||
| GenBank mRNA | AF198087.1 | - | 2 | 156 | ||
| AF453776.1 | - | 7 | 785 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmprss11d| Gene Symbol: Tmprss11d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033652.1 | NP_001028824.1 | 7 | 785 | ||
| NM_022630.1 | NP_072152.1 | 2 | 156 | |||
| GenBank mRNA | AF198087.1 | - | 2 | 156 | ||
| AF453776.1 | - | 7 | 785 | |||