siRNA Details
| Gene Symbol: Cul5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022683.1 | NP_073174.1 | 11 | 1274 | ||
| XM_006243090.2 | XP_006243152.1 | 10 | 1987 | |||
| GenBank mRNA | AF135115.1 | - | 11 | 1274 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001161618, NM_027807, AK031955, AK162082, AK014894, and others.

Cul5
Genomic Map