siRNA Details
| Gene Symbol: Sgta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240996.1 | XP_006241058.1 | 12 | 1840 | ||
| XM_006240997.1 | XP_006241059.1 | 12 | 1837 | |||
| XM_006240998.2 | XP_006241060.1 | 12 | 3680 | |||
| GenBank mRNA | AF368278.1 | - | 12 | 1747 | ||

Sgta
Genomic Map