siRNA Details
| Gene Symbol: Pln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022707.2 | NP_073198.1 | 2 | 658 | ||
| GenBank mRNA | CK356371.1 | - | 2 | 655 | ||
| X71068.1 | - | 2 | 656 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pln| Gene Symbol: Pln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022707.2 | NP_073198.1 | 2 | 658 | ||
| GenBank mRNA | CK356371.1 | - | 2 | 655 | ||
| X71068.1 | - | 2 | 656 | |||