siRNA Details
| Gene Symbol: BTN2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197233.2 | NP_001184162.1 | 4 | 1080 | ||
| NM_001197234.2 | NP_001184163.1 | 5 | 1192 | |||
| NM_007049.4 | NP_008980.1 | 5 | 1192 | |||
| NM_078476.3 | NP_510961.1 | 5 | 1192 | |||
| GenBank mRNA | AB209777.1 | - | 5 | 1128 | ||
| AK297098.1 | - | 4 | 1034 | |||
| BC016661.1 | - | 5 | 1135 | 4340392 | ||
| BX538050.1 | - | 4 | 735 | |||
| U90142.1 | - | 6 | 1110 | |||
| U90543.1 | - | 5 | 1133 | |||

BTN2A1
Genomic Map