siRNA Details
| Gene Symbol: Foxq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022858.2 | NP_074049.2 | 1 | 2019 | ||
| GenBank mRNA | L13201.1 | - | 1 | 1646 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxq1| Gene Symbol: Foxq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022858.2 | NP_074049.2 | 1 | 2019 | ||
| GenBank mRNA | L13201.1 | - | 1 | 1646 | ||