siRNA Details
| Gene Symbol: Map1lc3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022867.2 | NP_074058.2 | 3 | 266 | ||
| GenBank mRNA | AY206669.1 | - | 3 | 228 | ||
| AY310156.1 | - | 794 | ||||
| AY392036.1 | - | 3 | 266 | |||
| BC058144.1 | - | 3 | 224 | 6889616 | ||
| BC083556.1 | - | 3 | 246 | 7189855 | ||
| FQ221245.1 | - | 3 | 277 | |||
| FQ228912.1 | - | 3 | 277 | |||
| U05784.1 | - | 3 | 216 | |||

Map1lc3b
Genomic Map