siRNA Details
| Gene Symbol: Doc2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022937.2 | NP_075226.1 | 4 | 553 | ||
| XM_006230335.2 | XP_006230397.1 | 5 | 653 | |||
| XM_006230336.1 | XP_006230398.1 | 5 | 837 | |||
| XM_006230337.2 | XP_006230399.1 | 5 | 649 | |||
| XM_006230339.2 | XP_006230401.1 | 5 | 587 | |||
| XM_006230340.1 | XP_006230402.1 | 5 | 771 | |||
| XM_006230341.2 | XP_006230403.1 | 5 | 771 | |||
| XM_008759918.1 | XP_008758140.1 | 5 | 684 | |||

Doc2a
Genomic Map