siRNA Details
Gene Symbol: Pdzd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022940.1 | NP_075229.1 | 5683 | |||
GenBank mRNA | AF169411.1 | - | 5683 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pdzd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022940.1 | NP_075229.1 | 5683 | |||
GenBank mRNA | AF169411.1 | - | 5683 |