siRNA Details
| Gene Symbol: Dlgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767428.1 | XP_008765650.1 | 1 | 831 | ||
| GenBank mRNA | U67137.1 | - | 3 | 851 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dlgap1| Gene Symbol: Dlgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767428.1 | XP_008765650.1 | 1 | 831 | ||
| GenBank mRNA | U67137.1 | - | 3 | 851 | ||