siRNA Details
| Gene Symbol: Ppp1r10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022951.2 | NP_075240.1 | 4, 5 | 747 | ||
| XM_008772763.1 | XP_008770985.1 | 3, 4 | 1431 | |||
| XM_008772764.1 | XP_008770986.1 | 4, 5 | 1229 | |||
| GenBank mRNA | AF040954.1 | - | 3, 4 | 710 | ||
| BC134805.1 | - | 3, 4 | 720 | 8362313 | ||

Ppp1r10
Genomic Map