siRNA Details
Gene Symbol: Megf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022955.1 | NP_075244.1 | 4961 | |||
GenBank mRNA | AB011532.1 | - | 4961 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Megf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022955.1 | NP_075244.1 | 4961 | |||
GenBank mRNA | AB011532.1 | - | 4961 |