siRNA Details
| Gene Symbol: Dpysl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023023.1 | NP_075412.1 | 3 | 412 | ||
| GenBank mRNA | AB029432.1 | - | 2 | 289 | ||
| AJ131436.1 | - | 3 | 412 | |||
| Gene Symbol: LOC103692570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764540.1 | XP_008762762.1 | 523 | |||

Dpysl5
Genomic Map