siRNA Details
| Gene Symbol: Gucy1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023956.1 | NP_076446.1 | 4 | 1273 | ||
| GenBank mRNA | AB096080.1 | - | 4 | 1273 | ||
| AB097860.1 | - | 4 | 853 | |||
| AF109963.2 | - | 4 | 1273 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gucy1a2| Gene Symbol: Gucy1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023956.1 | NP_076446.1 | 4 | 1273 | ||
| GenBank mRNA | AB096080.1 | - | 4 | 1273 | ||
| AB097860.1 | - | 4 | 853 | |||
| AF109963.2 | - | 4 | 1273 | |||