siRNA Details
| Gene Symbol: Drp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023971.1 | NP_076461.1 | 4 | 506 | ||
| XM_006257235.2 | XP_006257297.1 | 3 | 477 | |||
| XM_008773432.1 | XP_008771654.1 | 4 | 633 | |||
| XM_008773433.1 | XP_008771655.1 | 4 | 389 | |||
| XM_008773434.1 | XP_008771656.1 | 4 | 604 | |||
| XM_008773435.1 | XP_008771657.1 | 3 | 477 | |||
| XM_008773436.1 | XP_008771658.1 | 2 | 1942 | |||
| GenBank mRNA | AF195787.1 | - | 4 | 506 | ||
| AF195788.1 | - | 3 | 439 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_010078, AK034383, AK038488, AK158102, BC125345, and others.

Drp2
Genomic Map