siRNA Details
| Gene Symbol: Drp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023971.1 | NP_076461.1 | 7, 8 | 1094 | ||
| XM_006257235.2 | XP_006257297.1 | 6, 7 | 1065 | |||
| XM_008773432.1 | XP_008771654.1 | 7, 8 | 1221 | |||
| XM_008773433.1 | XP_008771655.1 | 7, 8 | 977 | |||
| XM_008773434.1 | XP_008771656.1 | 7, 8 | 1192 | |||
| XM_008773435.1 | XP_008771657.1 | 6, 7 | 1065 | |||
| XM_008773436.1 | XP_008771658.1 | 5, 6 | 2530 | |||
| GenBank mRNA | AF195787.1 | - | 7, 8 | 1094 | ||
| AF195788.1 | - | 6, 7 | 1027 | |||

Drp2
Genomic Map