siRNA Details
| Gene Symbol: PKIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281444.1 | NP_001268373.1 | 6 | 1599 | ||
| NM_001281445.1 | NP_001268374.1 | 4 | 1162 | |||
| NM_007066.4 | NP_008997.1 | 4 | 1177 | |||
| NM_181804.2 | NP_861520.1 | 5 | 1324 | |||
| NM_181805.2 | NP_861521.1 | 6 | 1429 | |||
| XM_017027614.1 | XP_016883103.1 | 5 | 1308 | |||
| XM_017027615.1 | XP_016883104.1 | 4 | 1196 | |||
| XM_017027616.1 | XP_016883105.1 | 4 | 1114 | |||
| GenBank mRNA | AB019517.1 | - | 2 | 890 | ||
| AF115966.1 | - | 2 | 1139 | |||
| AF182032.1 | - | 4 | 1031 | |||
 Back To Top
 Back To Top
 PKIG
PKIG Genomic Map
Genomic Map Back To Top
 Back To Top