siRNA Details
| Gene Symbol: Camkv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024000.1 | NP_076490.1 | 2, 3 | 235 | ||
| GenBank mRNA | L22557.1 | - | 2, 3 | 235 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Camkv| Gene Symbol: Camkv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024000.1 | NP_076490.1 | 2, 3 | 235 | ||
| GenBank mRNA | L22557.1 | - | 2, 3 | 235 | ||